Dichlorobis(di-tert-butylphenylphosphine)palladium(II) (17,1% Pd)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Dichlorobis(di-tert-butylphenylphosphinE)palladium(ii)
45453.MD
45453.03
45453.MDEA
54.3
EUR
45453.MD
45453.03
Dichlorobis(di-tert-butylphenylphosphine)palladium(II) (17,1% Pd)
Dichlorobis(di-tert-butylphenylphosphine)palladium(II)
CAS No.: 34409-44-4
Molecular Formula: C28H46Cl2P2Pd
Formula Weight: 621.95
Physical Form: Solid
Sensitivity: Air and Moisture Sensitive
Appearance: Pale yellow
Melting Point: dec.
MDL No.: MFCD09953450
Formule:
C₂₈H₄₆Cl₂P₂Pd MW: 621.944 g/mol Smeltpunt: Dec. Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD09953450 CAS nummer: 34409-44-4 |