Iron-chrome based high temperature gas shift catalyst, HiFUEL® W210
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
45467.A1 45467.36 45467.A4
45467.A1EA 176 EUR
45467.A1 45467.36 45467.A4
Iron-chrome based high temperature gas shift catalyst, HiFUEL® W210
Iron-chrome based high temperature gas shift catalyst
Physical Form: Pellets, 5.4mm x 3.6mm Appearance: Red-brown