Barium dodecairon nonadecaoxide
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
45552.14
45552.22
45552.14EA
90.2
EUR
45552.14
45552.22
Barium dodecairon nonadecaoxide
Barium dodecairon nonadecaoxide
CAS No.: 12047-11-9
Molecular Formula: BaFe12O19
Formula Weight: 1111.48
Physical Form: Powder
Appearance: Black
Melting Point:
Formule:
BaFe₁₂O₁₉ Smeltpunt: >1315 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00075637 CAS nummer: 12047-11-9 EINECS: 234-974-5 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...