Nitinol (alloy M), tension coil spring wire diameter 0.75 mm (0.030 in), mean coil diameter 5.55 mm (0.22 in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
45655.KJ
45655.KM
45655.KJEA
83.7
EUR
45655.KJ
45655.KM
Nitinol (alloy M), tension coil spring wire diameter 0.75 mm (0.030 in), mean coil diameter 5.55 mm (0.22 in)
Nitinol (alloy M)
Odour: Odourless
MDL No.: MFCD02091734