Palladium 5% on activated carbon paste (50% water) 5R394
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45921.06 45921.14
45921.06EA 82.7 EUR
45921.06 45921.14
Palladium 5% on activated carbon paste (50% water) 5R394
Palladium
CAS No.: 7440-05-3 Density: 1.5 Appearance: Black Odour: Odourless MDL No.: MFCD03457879
Formule: Pd
MW: 106.420 g/mol
Dichtheid: 1.5
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6