Platinum 5% on activated carbon wetted with ca. 50% water, powder, reduced Type B105022-5
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Platinium
46306.06
46306.14
46306.22
46306.06EA
101
EUR
46306.06
46306.14
46306.22
Platinum 5% on activated carbon wetted with ca. 50% water, powder, reduced Type B105022-5
Platinum
CAS No.: 7440-06-4
Physical Form: 900-1100 m²/g
Appearance: Dark grey to black
Odour: Odourless
MDL No.: MFCD00011179
Bestelinformatie: Sold on a dry weight basis. Unit weight excludes water weight.
Opgelet: Incompatible with acids and oxidising agents
Formule:
Pt Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011179 CAS nummer: 7440-06-4 |