Tetrabutylammonium hydroxide 1.0 M in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
46315.AE 46315.AP
46315.AEEA 147 EUR
46315.AE 46315.AP
Tetrabutylammonium hydroxide 1.0 M in aqueous solution
Tetrabutylammonium hydroxide
CAS No.: 2052-49-5 Molecular Formula: C16H37NO Formula Weight: 259.48 Flash Point: None Physical Form: Liquid Sensitivity: Air Sensitive UN No.: UN3267 MDL No.: MFCD00009425
Formule: C₁₆H₃₇NO
MW: 259.48 g/mol
Smeltpunt: 29.85
Dichtheid: 0.968
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II