Inconel® 600 alloy, gauze_wire 10 mesh woven from 0.64mm (0.025in)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
46453.FL
46453.GT
46453.HB
46453.FLEA
27
EUR
46453.FL
46453.GT
46453.HB
Inconel® 600 alloy, gauze_wire 10 mesh woven from 0.64mm (0.025in)
Inconel® 600 alloy
Molecular Formula: Ni:Cr:Fe; 77:15.5:7.5 wt%
Physical Form: Wire Cloth
MDL No.: MFCD01741927