Antimony, plasma standard solution, Sb 1000 µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
47103.AD
47103.AE
47103.ADEA
59.2
EUR
47103.AD
47103.AE
Antimony, plasma standard solution, Sb 1000 µg/ml, Specpure®
Standaarden
Kationische standaarden
Antimonium standaarden
Molecular Formula: Sb in 30% HCl
Physical Form: Liquid
Odour: Acidic
UN No.: UN1789
MDL No.: MFCD00134030