Antimony, AAS standard solution, Sb 1000 µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
47197.AE 47197.AP
47197.AEEA 52.2 EUR
47197.AE 47197.AP
Antimony, AAS standard solution, Sb 1000 µg/ml, Specpure®
Standaarden Kationische standaarden Antimonium standaarden
Molecular Formula: Sb in 30% HCl
Physical Form: Liquid
Odour: Acidic
UN No.: UN1789
MDL No.: MFCD00134030