Iron, plasma standard solution, Fe 1000 µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
47206.AD 47206.AE 47206.AP
47206.ADEA 65.2 EUR
47206.AD 47206.AE 47206.AP
Iron, plasma standard solution, Fe 1000 µg/ml, Specpure®
Standaarden Kationische standaarden IJzer standaarden
Molecular Formula: (Fe(NO₃)₃).9H₂O in 5% HNO₃
UN No.: UN3265
MDL No.: MFCD00010999
Order Now
Specificaties
Environmentally Preferable
MDL No. MFCD00010999