Iron, plasma standard solution, Fe 10000 µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
47221.AD 47221.AE 47221.AP
47221.ADEA 95.1 EUR
47221.AD 47221.AE 47221.AP
Iron, plasma standard solution, Fe 10000 µg/ml, Specpure®
Standaarden Kationische standaarden IJzer standaarden
Molecular Formula: (Fe(NO₃)₃).9H₂O in 5% HNO₃
Physical Form: Liquid
Odour: Acidic
UN No.: UN3265
MDL No.: MFCD00010999