Iron, AAS standard solution, Fe 1000 µg/ml, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
47282.AE
47282.AP
47282.AEEA
49.6
EUR
47282.AE
47282.AP
Iron, AAS standard solution, Fe 1000 µg/ml, Specpure®
Standaarden
Kationische standaarden
IJzer standaarden
Molecular Formula: (Fe(NO₃)₃).9H₂O in 5% HNO₃
Physical Form: Liquid
Odour: Acidic
UN No.: UN3265
MDL No.: MFCD00010999