Cerium iron oxide (50:50 wt%) 20% in water, nanoparticle dispersion <5.0 nm APS NW, low pH
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
47283.22
47283.36
47283.22EA
218
EUR
47283.22
47283.36
Cerium iron oxide (50:50 wt%) 20% in water, nanoparticle dispersion <5.0 nm APS NW, low pH
Cerium iron oxide
Molecular Formula: CeO2:Fe2O3; =50:50 wt%
Physical Form: Liquid
Density: 1.23
Appearance: Red-brown
Odour: Petroleum-like
MDL No.: MFCD02687051