Copper phosphorus shot
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88357.36
88357.A3
88357.36EA
158
EUR
88357.36
88357.A3
Copper phosphorus shot
Copper phosphorus shot
Molecular Formula: Cu:P; 85:15 wt%
MDL No.: MFCD01074956
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...