Titanium nickel composite (70:30), powder -325 mesh
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
88384.22
88384.36
88384.22EA
232
EUR
88384.22
88384.36
Titanium nickel composite (70:30), powder -325 mesh
Titanium nickel composite
Molecular Formula: Ti:Ni; 70:30 wt%
Sensitivity: Air Sensitive
UN No.: UN3089
MDL No.: MFCD02091734
Formule:
Ti:Ni Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD02091734 UN: 3089 ADR: 4.1,II |