Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -325 mesh Type 316-L
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
88390.22
88390.36
88390.A3
88390.22EA
67.5
EUR
88390.22
88390.36
88390.A3
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -325 mesh Type 316-L
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Appearance: Gray
MDL No.: MFCD00148499