Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) ≥75%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Diisopropoxytitanium(IV) Bis(acetylacetonate), Titanium (di-i-propoxide)bis(acetylacetonate), Titanium(IV) Diisopropoxide Bis(acetylacetonate), Titanium (IV) bis (acetylacetonate) diisopropoxide
89360.22
89360.36
89360.22EA
40
EUR
89360.22
89360.36
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) ≥75%
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV)
CAS No.: 17927-72-9
Molecular Formula: C16H28O6Ti
Formula Weight: 364.30
Flash Point: 18°(61°F)
Physical Form: Liquid
Density: 1.010
Refractive Index: 1.494
Sensitivity: Moisture Sensitive
Appearance: Brown
Odour: Alcohol-like
UN No.: UN1219
MDL No.: MFCD00000031
Formule:
C₁₆H₂₈O₆Ti MW: 364.26 g/mol Dichtheid: 1.010 Vlampunt: 18 °C (61 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000031 CAS nummer: 17927-72-9 EINECS: 241-866-1 UN: 1219 ADR: 3 II |