Dichloromethylenedimethyliminium chloride 95%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 95%
CAS No.: 33842-02-3
Molecular Formula: C3H6Cl3N
Formula Weight: 162.45
Storage Temperature: 2°C to 8°C
Melting Point: ca 185° dec.
Beilstein Reference: 4157987
UN No.: UN3261
MDL No.: MFCD00011870
Formule:
C₃H₆Cl₃N |
MDL Nummer:
MFCD00011870 CAS nummer: 33842-02-3 EINECS: 251-695-4 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...