L-Valine methyl ester hydrochloride 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
H-Val-OMe.HCl, L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, L-Methyl valinate hydrochloride, H-Val-OMe·HCl
A15892.09
A15892.18
A15892.09EA
60.6
EUR
A15892.09
A15892.18
L-Valine methyl ester hydrochloride 99%
L-Valine methyl ester hydrochloride
Purity: 99%
CAS No.: 6306-52-1
Molecular Formula: C6H13NO2.HCl
Formula Weight: 167.64
Storage Temperature: 2°C to 8°C
Melting Point: 166° dec.
Beilstein Reference: 3594960
Optical Rotation: +16° (c=2 in water)
MDL No.: MFCD00012497
Formule:
C₆H₁₄ClNO₂ MW: 167.63 g/mol Smeltpunt: 166 °C (ontbinding) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00012497 CAS nummer: 6306-52-1 EINECS: 228-620-9 |