Chlorophyllin, coppered trisodium salt
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A16155.06 A16155.14 A16155.22
A16155.06EA 34.3 EUR
A16155.06 A16155.14 A16155.22
Chlorophyllin, coppered trisodium salt
Chlorophyllin, coppered trisodium salt
CAS No.: 11006-34-1 Molecular Formula: C34H31CuN4Na3O6 Formula Weight: 724.15 Storage Temperature: 2°C to 8°C Odour: Odourless MDL No.: MFCD00012149
Formule: C₃₄H₃₁CuN₄Na₃O₆
MW: 724.15 g/mol
MDL Nummer: MFCD00012149
CAS nummer: 11006-34-1
EINECS: 234-242-5