3-Hepten-2-one 97%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 97%
CAS No.: 1119-44-4
Molecular Formula: C7H12O
Formula Weight: 112.16
Storage Temperature: 2°C to 8°C
Flash Point: 49°(120°F)
Refractive Index: 1.4450
Boiling Point: 156°
Beilstein Reference: 1739239
UN No.: UN1224
MDL No.: MFCD00015564
Formule:
C₇H₁₂O Kookpunt: 156 °C Dichtheid: 0,860 Vlampunt: 49 °C (120 °F) |
MDL Nummer:
MFCD00015564 CAS nummer: 1119-44-4 EINECS: 214-278-8 ADR: 3,III |