S-Methyl thiobutyrate 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Thiobutyric Acid S-Methyl Ester
B22144.06
B22144.14
B22144.06EA
44.7
EUR
B22144.06
B22144.14
S-Methyl thiobutyrate 98%
S-Methyl thiobutyrate
Purity: 98%
CAS No.: 2432-51-1
Molecular Formula: C5H10OS
Formula Weight: 118.20
Flash Point: 34°(93°F)
Refractive Index: 1.4620
Boiling Point: 142-143°
Beilstein Reference: 1848987
UN No.: UN1993
MDL No.: MFCD00009872
Formule:
C₅H₁₀OS Kookpunt: 142…143 °C Dichtheid: 0,966 Vlampunt: 34 °C (93 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00009872 CAS nummer: 2432-51-1 EINECS: 219-407-1 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...