Methallyl phenyl ether 96%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B22369.06
B22369.14
B22369.06EA
40.8
EUR
B22369.06
B22369.14
Methallyl phenyl ether 96%
Methallyl phenyl ether
Purity: 96%
CAS No.: 5820-22-4
Molecular Formula: C10H12O
Formula Weight: 148.21
Flash Point: 77°(171°F)
Refractive Index: 1.5150
Boiling Point: 204°
Beilstein Reference: 1933868
MDL No.: MFCD00008597
Formule:
C₁₀H₁₂O |
MDL Nummer:
MFCD00008597 CAS nummer: 5820-22-4 |