DL-α-Amino-2-thiopheneacetic acid methyl ester hydrochloride 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
B25190.03
B25190.06
B25190.03EA
59.2
EUR
B25190.03
B25190.06
DL-α-Amino-2-thiopheneacetic acid methyl ester hydrochloride 98%
DL-alpha-Amino-2-thiopheneacetic acid methyl ester hydrochloride
Purity: 98%
Molecular Formula: C7H9NO2S.HCl
Formula Weight: 207.67
Melting Point: ca 180° dec.
MDL No.: MFCD04039779