1-Benzyl-1,4,7,10-tetraazacyclododecane
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H26525.MD
H26525.03
H26525.MDEA
132
EUR
H26525.MD
H26525.03
1-Benzyl-1,4,7,10-tetraazacyclododecane
1-Benzyl-1,4,7,10-tetraazacyclododecane
CAS No.: 112193-83-6
Molecular Formula: C15H26N4
Formula Weight: 262.40
Melting Point: 84-86°
MDL No.: MFCD09263310
Formule:
C₁₅H₂₆N₄ Smeltpunt: 84…86 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD09263310 CAS nummer: 112193-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...