L-Serine benzyl ester hydrochloride 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
H-Ser-OBzl·HCl, H-Ser-OBzl.HCl
H28790.14
H28790.06
H28790.03
H28790.14EA
395
EUR
H28790.14
H28790.06
H28790.03
L-Serine benzyl ester hydrochloride 98%
L-Serine benzyl ester hydrochloride
Purity: 98%
CAS No.: 60022-62-0
Molecular Formula: C10H13NO3.HCl
Formula Weight: 231.68
Storage Temperature: 2°C to 8°C
Melting Point: ca 175° dec.
Optical Rotation: -12° (c=4 in water)
MDL No.: MFCD00038955
Formule:
C₁₀H₁₃NO₃·HCl MW: 231.6 g/mol Smeltpunt: ∼ 175 °C (ontbinding) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00038955 CAS nummer: 60022-62-0 |