2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H31846.MD
H31846.03
H31846.MDEA
119
EUR
H31846.MD
H31846.03
2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole 95%
2-Iodo-1-(2-tetrahydropyranyl)-1H-imidazole
Purity: 95%
Molecular Formula: C8H11IN2O
Formula Weight: 278.09
MDL No.: MFCD11100955