4-Isopropylbenzenesulphonamide 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H32950.03
H32950.06
H32950.03EA
56
EUR
H32950.03
H32950.06
4-Isopropylbenzenesulphonamide 97%
4-Isopropylbenzenesulphonamide
Purity: 97%
CAS No.: 6335-39-3
Molecular Formula: C9H13NO2S
Formula Weight: 199.27
Melting Point: 103-106°
MDL No.: MFCD00457071
Formule:
C₉H₁₃NO₂S MW: 199.27 g/mol Smeltpunt: 103…106 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00457071 CAS nummer: 6335-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...