(E)-1-Ethoxyethene-2-boronic acid pinacolester 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H33367.03
H33367.06
H33367.03EA
144
EUR
H33367.03
H33367.06
(E)-1-Ethoxyethene-2-boronic acid pinacolester 98%
(E)-1-Ethoxyethene-2-boronic acid pinacolester
Purity: 98%
CAS No.: 1201905-61-4
Molecular Formula: C10H19BO3
Formula Weight: 198.07
Refractive Index: 1.4460
Boiling Point: 50°/0.3mm
MDL No.: MFCD09702441
Formule:
C₁₀H₁₉BO₃ Kookpunt: 50 °C (0,3 mmHg) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD09998813 CAS nummer: 1201905-61-4 |