Dicyclohexylacetic acid ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H33523.06
H33523.14
H33523.06EA
77.6
EUR
H33523.06
H33523.14
Dicyclohexylacetic acid ≥98%
Dicyclohexylacetic acid
Purity: 98+%
CAS No.: 52034-92-1
Molecular Formula: C14H24O2
Formula Weight: 224.34
Melting Point: 139-141°
MDL No.: MFCD00075017
Formule:
C₁₄H₂₄O₂ Smeltpunt: 139…141 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00075017 CAS nummer: 52034-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...