Platinum 5% on carbon paste wetted with ca. 50% water, sulfided B106032-5
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Platinium
H36544.06 H36544.14
H36544.06EA 96.6 EUR
H36544.06 H36544.14
Platinum 5% on carbon paste wetted with ca. 50% water, sulfided B106032-5
Platinum
CAS No.: 7440-06-4 Physical Form: Solid Appearance: Dark grey to black Odour: Odourless MDL No.: MFCD00011179
Formule: Pt
MW: 195.08 g/mol
Kookpunt: 3827
Smeltpunt: 1769
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689