4-(1-Pyrrolidinyl)-1-butylamine 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
4-(Pyrrolidin-1-yl)butan-1-amine
H50323.03
H50323.06
H50323.03EA
56.7
EUR
H50323.03
H50323.06
4-(1-Pyrrolidinyl)-1-butylamine 98%
4-(1-Pyrrolidinyl)-1-butylamine
Purity: 98%
CAS No.: 24715-90-0
Molecular Formula: C8H18N2
Formula Weight: 142.25
UN No.: UN2735
MDL No.: MFCD00192591
Formule:
C₈H₁₈N₂ MW: 142.25 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00192591 CAS nummer: 24715-90-0 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...