5-Chlorothiophene-2-carbonyl chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
H61474.06
H61474.14
H61474.06EA
119
EUR
H61474.06
H61474.14
5-Chlorothiophene-2-carbonyl chloride 98%
5-Chlorothiophene-2-carbonyl chloride
Purity: 98%
CAS No.: 42518-98-9
Molecular Formula: C5H2Cl2OS
Formula Weight: 181.04
Storage Temperature: 2°C to 8°C
Melting Point: 4°
UN No.: UN3265
MDL No.: MFCD00043886
Formule:
C5H2Cl2OS MW: 181.04 g/mol |
MDL Nummer:
MFCD00043886 CAS nummer: 42518-98-9 ADR: 8 II |