1-(Triphenylmethyl)-L-histidine 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
H-His(Trt)-OH, 1-Trityl-L-histidine, H-His(1-Trt)-OH
H62779.03
H62779.06
H62779.03EA
58
EUR
H62779.03
H62779.06
1-(Triphenylmethyl)-L-histidine 98%
1-(Triphenylmethyl)-L-histidine
Purity: 98%
CAS No.: 35146-32-8
Molecular Formula: C25H23N3O2
Formula Weight: 397.47
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 210° dec
Beilstein Reference: 767244
MDL No.: MFCD00153444
Formule:
C₂₅H₂₃N₃O₂ MW: 397.478 g/mol Smeltpunt: 210 °C (ontbinding) |
MDL Nummer:
MFCD00153444 CAS nummer: 35146-32-8 |