4-Aminocarbethoxypiperidine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

H63095.03 H63095.06
H63095.03EA 85.1 EUR
H63095.03 H63095.06
4-Aminocarbethoxypiperidine 97%
4-Aminocarbethoxypiperidine

Purity: 97% CAS No.: 64951-36-6 Molecular Formula: C8H16N2O2 Formula Weight: 172.22 Physical Form: Solid MDL No.: MFCD03844679

Formule: C₈H₁₆N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03844679
CAS nummer: 64951-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR