2-Bromo-L-phenylalanine 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H63141.03
H63141.06
H63141.03EA
102
EUR
H63141.03
H63141.06
2-Bromo-L-phenylalanine 95%
2-Bromo-L-phenylalanine
Purity: 95%
CAS No.: 42538-40-9
Molecular Formula: C9H10BrNO2
Formula Weight: 244.09
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD03092958
Formule:
C₉H₁₀BrNO₂ |
MDL Nummer:
MFCD03092958 CAS nummer: 42538-40-9 |