L-Tryptophanamide hydrochloride 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H63405.03
H63405.06
H63405.03EA
40.2
EUR
H63405.03
H63405.06
L-Tryptophanamide hydrochloride 95%
L-Tryptophanamide hydrochloride
Purity: 95%
CAS No.: 5022-65-1
Molecular Formula: C11H13N3O.HCl
Formula Weight: 239.70
Physical Form: Solid
MDL No.: MFCD00054315
Formule:
C₁₁H₁₃N₃O |
MDL Nummer:
MFCD00054315 CAS nummer: 5022-65-1 |