Fmoc-Dap(Alloc)-OH 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Fmoc-Dap(Aloc)-OH, (S)-3-Allyloxycarbonylamino-2-(Fmoc-amino)propionic acid
H63516.03
H63516.06
H63516.03EA
164
EUR
H63516.03
H63516.06
Fmoc-Dap(Alloc)-OH 95%
Fmoc-Dap(Alloc)-OH
Purity: 95%
CAS No.: 188970-92-5
Molecular Formula: C22H22N2O6
Formula Weight: 410.42
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 9161065
MDL No.: MFCD00273468
Formule:
C₂₂H₂₂N₂O₆ |
MDL Nummer:
MFCD00273468 CAS nummer: 188970-92-5 |