Nalpha-(tert-Butoxycarbonyl)-L-arginine hydrochloride 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Nalpha-(tert-Butoxycarbonyl)-L-arginine hydrochloride, Boc-Arg-OH.HCl
H63611.06
H63611.14
H63611.06EA
35.8
EUR
H63611.06
H63611.14
Nalpha-(tert-Butoxycarbonyl)-L-arginine hydrochloride 98%
Nalpha-Boc-L-arginine hydrochloride
Purity: 98%
CAS No.: 35897-34-8
Molecular Formula: C11H22N4O4.HCl
Formula Weight: 310.78
Physical Form: Powder
Appearance: White
Beilstein Reference: 3658939
MDL No.: MFCD00063594
Formule:
C₁₁H₂₂N₄O₄·HCl MW: 310.78 g/mol Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00063594 CAS nummer: 35897-34-8 |