H-Tyr(BZL)-OMe·HCl 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H63908.03
H63908.06
H63908.03EA
28.8
EUR
H63908.03
H63908.06
H-Tyr(BZL)-OMe·HCl 98%
H-Tyr(BZL)-OMe·HCl
Purity: 98%
CAS No.: 34805-17-9
Molecular Formula: C17H19NO3.HCl
Formula Weight: 321.8
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00080888
Formule:
C₁₇H₂₀ClNO₃ |
MDL Nummer:
MFCD00080888 CAS nummer: 34805-17-9 |