2-Bromo-3-fluorobenzaldehyde 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

H64400.06 H64400.14 H64400.22
H64400.06EA 65 EUR
H64400.06 H64400.14 H64400.22
2-Bromo-3-fluorobenzaldehyde 98%
2-Bromo-3-fluorobenzaldehyde

Purity: 98% CAS No.: 891180-59-9 Molecular Formula: C7H4BrFO Formula Weight: 203.01 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Light Sensitive Beilstein Reference: 10529128 MDL No.: MFCD07782044

Formule: C₇H₄BrFO
MDL Nummer: MFCD07782044
CAS nummer: 891180-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR