4-Bromo-1-methyl-1H-pyrazole 98+%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 98+%
CAS No.: 15803-02-8
Molecular Formula: C4H5BrN2
Formula Weight: 161.00
Flash Point: 94°(201°F)
Physical Form: Liquid
Density: 1.558
Refractive Index: 1.5310
Appearance: Colourless to pale yellow
Boiling Point: 185-188°
Beilstein Reference: 108120
MDL No.: MFCD02179565
Formule:
C₄H₅BrN₂ Kookpunt: 185…188 °C Dichtheid: 1,558 Vlampunt: 94 °C (201 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD02179565 CAS nummer: 15803-02-8 |