5-Bromo-2-chloro-3-nitropyridine 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H64897.14
H64897.22
H64897.14EA
64.3
EUR
H64897.14
H64897.22
5-Bromo-2-chloro-3-nitropyridine 98%
5-Bromo-2-chloro-3-nitropyridine
Purity: 98%
CAS No.: 67443-38-3
Molecular Formula: C5H2BrClN2O2
Formula Weight: 237.44
Physical Form: Crystalline
Appearance: Yellow
Melting Point: 66-68°
Beilstein Reference: 147551
UN No.: UN2811
MDL No.: MFCD00222270
Formule:
C₅H₂BrClN₂O₂ MW: 237.44 g/mol Smeltpunt: 66…68 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00222270 CAS nummer: 67443-38-3 ADR: 6.1 III |