Sodium orthovanadate 100 mM in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
J60191.AD
J60191.AE
J60191.ADEA
72.9
EUR
J60191.AD
J60191.AE
Sodium orthovanadate 100 mM in aqueous solution
Sodium orthovanadate
CAS No.: 13721-39-6
Molecular Formula: Na3O4V
Formula Weight: 183.94
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00003511
Formule:
Na₃O₄V Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00003511 CAS nummer: 13721-39-6 |