EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M in aqueous solution pH 8.0
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI
J60767.ADEA 100 EUR
J60767.AD J60767.AE
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M in aqueous solution pH 8.0
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)
CAS No.: 67-42-5 Molecular Formula: C14H24N2O10 Formula Weight: 380.35 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00004291
Formule: C₁₄H₂₄N₂O₁₀
MW: 380.35 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004291
CAS nummer: 67-42-5
Merck Index: 13,03560