Agarose gel loading dye 6X for electrophoresis, glycerol based, no xylene Cyanol FF
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J60933.AC
J60933.AD
J60933.ACEA
67.5
EUR
J60933.AC
J60933.AD
Agarose gel loading dye 6X for electrophoresis, glycerol based, no xylene Cyanol FF
Kleurstoffen
Kleuringen voor elektroforese
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels.
- Glycerol based
- Xylene cyanol FF free
- 0,25 % bromophenol blue and 30 % glycerol
Storage Temperature: 2°C to 8°C
Physical Form: Liquid