Laemmli SDS sample buffer with pyronin Y, reducing (4X)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J62115.AC J62115.AD
J62115.ACEA 54.3 EUR
J62115.AC J62115.AD
Laemmli SDS sample buffer with pyronin Y, reducing (4X)
Buffers Buffers voor elektroforese Loading-buffers
Physical Form: Liquid
Appearance: Pink
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system