Nicergoline ≥95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

J63295.MC J63295.03
J63295.MCEA 56.2 EUR
J63295.MC J63295.03
Nicergoline ≥95%
Nicergoline

CAS No.: 27848-84-6 Molecular Formula: C24H26BrN3O3 Formula Weight: 484.39 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00869626


Applications: Cerebral and peripheral vasodilator

Formule: C₂₄H₂₆BrN₃O₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00869626
CAS nummer: 27848-84-6
EINECS: 248-694-6
Merck Index: 14,6496

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR