Laemmli SDS sample buffer, non-reducing (4X)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J63615.AC J63615.AD
J63615.ACEA 61.9 EUR
J63615.AC J63615.AD
Laemmli SDS sample buffer, non-reducing (4X)
Buffers Buffers voor elektroforese Loading-buffers
Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system